missing translation for 'onlineSavingsMsg'
Learn More

VPS33B Antibody - BSA Free, Novus Biologicals™

Product Code. 30558219 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30558219 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30558219 Supplier Novus Biologicals Supplier No. NBP345589

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

VPS33B Polyclonal antibody specifically detects VPS33B in Human,Mouse,Rat,Zebrafish samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS33B
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:10-1:100
Formulation PBS, 50% Glycerol, pH (7.3)
Gene Alias FLJ14848, hVPS33B, vacuolar protein sorting 33 homolog B (yeast), vacuolar protein sorting 33B, vacuolar protein sorting 33B (yeast homolog), vacuolar protein sorting-associated protein 33B
Host Species Rabbit
Immunogen vacuolar protein sorting 33 homolog B(yeast). (Uniprot# Q9H267)
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 26276
Target Species Human, Mouse, Rat, Zebrafish
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.