missing translation for 'onlineSavingsMsg'
Learn More

vWF-A2 Antibody (7G2B11C10), Novus Biologicals™

Product Code. p-200058915 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18602718 100 μg 100µL
18692799 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18602718 Supplier Novus Biologicals Supplier No. NBP306526100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

vWF-A2 Monoclonal antibody specifically detects vWF-A2 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen vWF-A2
Applications ELISA
Classification Monoclonal
Clone 7G2B11C10
Conjugate Unconjugated
Dilution ELISA 1:2000-1:4000
Formulation 0.2 um filtered solution in PBS
Gene Alias F8VWF, VWD, VWF von Willebrand factor
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human vWF-A2 (Accession#: AAB59458.1; Met1-Lys2813).
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7450
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.