missing translation for 'onlineSavingsMsg'
Learn More

Warfarin EP Impurity B, Mikromol™

Product Code. 30462816
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30462816 4 x 25 mg -
30050796 25 mg 25mg
17923136 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30462816 Supplier Mikromol Supplier No. MM0732.0200254AMP
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Warfarin EP Impurity B, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxycoumarin
CAS 1076-38-6
Analyte or Component Names 4-Hydroxycoumarin
Shipping Condition Room Temperature
Molecular Formula C9 H6 O3
Certifications/Compliance ISO 17025
Synonym 4-Hydroxy-2H-1-benzopyran-2-one (4-Hydroxycoumarin), Warfarin Sodium Imp. B (EP), Warfarin Imp. B (EP), 4-Hydroxy-2H-1-benzopyran-2-one, 4-Hydroxycoumarin, 4-Coumarinol, 4-Hydroxy-2H-benzo[b]pyran-2-one, 4-Hydroxychromen-2-one, Benzotetronic acid, NSC 11889, Warfarin Sodium Impurity B, Warfarin Impurity B
InChI Formula InChI=1S/C9H6O3/c10-7-5-9(11)12-8-4-2-1-3-6(7)8/h1-5,10H
Solution Type Neat
SMILES OC1=CC(=O)Oc2ccccc12
IUPAC Name 4-hydroxychromen-2-one
Molecular Weight (g/mol) 162.14
Quantity 4 x 25 mg
Formula Weight 162.0317 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.