missing translation for 'onlineSavingsMsg'
Learn More

Warfarin, Lipomed

Product Code. 30460215
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460215 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460215 Supplier Lipomed Supplier No. LPMWAR192210
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Warfarin
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Warfarin
CAS 81-81-2
Analyte or Component Names Warfarin
Shipping Condition Room Temperature
Molecular Formula C19 H16 O4
Synonym 2 H-1-Benzopyran-2-one, 4-hydroxy-3-(3-oxo-1-phenylbutyl)-, 4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-1-benzopyran-2-one, (RS)-Warfarin, 1-(4'-Hydroxy-3'-coumarinyl)-1-phenyl-3-butanone, 3-(1'-Phenyl-2'-acetylethyl)-4-hydroxycoumarin, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2 H-chromen-2-one
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES CC(=O)CC(C1=C(O)c2ccccc2OC1=O)c3ccccc3
IUPAC Name 4-hydroxy-3-(3-oxo-1-phenylbutyl)chromen-2-one
Industry Type Pharmaceutical
Molecular Weight (g/mol) 308.33
Quantity 10 mg
Formula Weight 308.1049 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.