missing translation for 'onlineSavingsMsg'
Learn More

Warfarin, Mikromol™

Product Code. 18091100
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18091100 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18091100 Supplier Mikromol Supplier No. MM0638.00
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Warfarin
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Warfarin
CAS 81-81-2
Analyte or Component Names Warfarin
Shipping Condition Room Temperature
Molecular Formula C19 H16 O4
Certifications/Compliance ISO 17034
Synonym 2H-1-Benzopyran-2-one, 4-hydroxy-3-(3-oxo-1-phenylbutyl)-, 4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2H-1-benzopyran-2-one, (RS)-Warfarin, 1-(4'-Hydroxy-3'-coumarinyl)-1-phenyl-3-butanone, 3-(1'-Phenyl-2'-acetylethyl)-4-hydroxycoumarin, 4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2H-chromen-2-one, Athrombine-K, Brumolin, Circuvit, Co-Rax, Compound 42, Coumafen, Coumafene, Coumaphen, Coumefene, Dethmor, Kumader, Kumadu, Kumatox, Martefarin, NSC 59813, Ratron, Ratron G, Rodafarin, Rodafarin C, Rodex, Temus W, Vampirinip II, Vampirinip III, W.A.R.F. 42, WARF compound 42, Warf 5, Warfarin, Warfarin acid, Zoocoumarin, rac-Warfarin, Coumarin, 3-(alpha-acetonylbenzyl)-4-hydroxy- (7CI,8CI), (+/-)-Warfarin, (+/-)-Warfarin-alcohol, 3-(alpha-Acetonylbenzyl)-4-hydroxycoumarin, 3-(alpha-Phenyl-beta-acetylethyl)-4-hydroxycoumarin, DL-3-(alpha-Acetonylbenzyl)-4-hydroxycoumarin, (RS)-4-Hydroxy-3-(3-oxo-1-phenylbutyl)-2H-chromen-2-one
InChI Formula InChI=1S/C19H16O4/c1-12(20)11-15(13-7-3-2-4-8-13)17-18(21)14-9-5-6-10-16(14)23-19(17)22/h2-10,15,21H,11H2,1H3
SMILES CC(=O)CC(C1=C(O)c2ccccc2OC1=O)c3ccccc3
IUPAC Name 4-hydroxy-3-(3-oxo-1-phenylbutyl)chromen-2-one
Molecular Weight (g/mol) 308.33
Quantity 100 mg
Formula Weight 308.1049 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.