missing translation for 'onlineSavingsMsg'
Learn More
Learn More
West Nile Virus Envelope Antibody, mFluor Violet 610 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
West Nile Virus Envelope Polyclonal antibody specifically detects West Nile Virus Envelope in Virus samples. It is validated for ELISA
Specifications
Specifications
| Antigen | West Nile Virus Envelope |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 610 SE |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a synthetic peptide corresponding to 14 amino acids at the C-terminus of the West Nile Virus Envelope. The immunogen is located within amino acids 740 - 790 of West Nile Virus Envelope Protein. Amino Acid Squence: MGINARDRSIAMTF |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?