missing translation for 'onlineSavingsMsg'
Learn More

Wnt-5a Antibody (CL13683) [Allophycocyanin], Novus Biologicals™

Product Code. 30525465 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30525465 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30525465 Supplier Novus Biologicals Supplier No. NBP343867APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Wnt-5a Monoclonal antibody specifically detects Wnt-5a in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Wnt-5a
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL13683
Conjugate APC
Formulation PBS
Gene Alias hWNT5A, protein Wnt-5a, wingless-type MMTV integration site family, member 5A, WNT-5A protein
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of P41221, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer Stem Cells, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 7474
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.