missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wnt-5b Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41306
This item is not returnable.
View return policy
Description
Wnt-5b Polyclonal specifically detects Wnt-5b in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Wnt-5b | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| WNT5B | |
| Antibody was raised against a 15 amino acid peptide near the center of human WNT5B. The immunogen is located within amino acids 130 - 180 of WNT5. Amino Acid Squence: CSRTARPKDLPRDWL | |
| Affinity Purified | |
| RUO | |
| Primary | |
| WNT5 antibody will detect both WNT5A and WNT5B. At least two isoforms of WNT5B are known to exist; this antibody will detect both isoforms. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin 1:10-1:500 | |
| MGC2648, protein Wnt-5b, wingless-type MMTV integration site family, member 5B, WNT-5B protein | |
| Rabbit | |
| 465 kDa | |
| 0.1 mg | |
| Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| 81029 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction