missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wnt-5b Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
184.00 € - 507.00 €
Specifications
| Antigen | Wnt-5b |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18678348
|
Novus Biologicals
NBP2-41306-0.025mg |
0.025 mg |
184.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18170639
|
Novus Biologicals
NBP2-41306 |
0.1 mg |
507.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Wnt-5b Polyclonal specifically detects Wnt-5b in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Wnt-5b | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| MGC2648, protein Wnt-5b, wingless-type MMTV integration site family, member 5B, WNT-5B protein | |
| WNT5B | |
| IgG | |
| Affinity Purified | |
| 465 kDa |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| PBS with 0.02% Sodium Azide | |
| 81029 | |
| Antibody was raised against a 15 amino acid peptide near the center of human WNT5B. The immunogen is located within amino acids 130 - 180 of WNT5. Amino Acid Squence: CSRTARPKDLPRDWL | |
| Primary | |
| WNT5 antibody will detect both WNT5A and WNT5B. At least two isoforms of WNT5B are known to exist; this antibody will detect both isoforms. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title