missing translation for 'onlineSavingsMsg'
Learn More

WRN Antibody (SR1313), Novus Biologicals™

Product Code. p-200005630 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18661765 100 μg 100µL
18659434 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18661765 Supplier Novus Biologicals Supplier No. NBP322281100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

WRN Monoclonal antibody specifically detects WRN in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen WRN
Applications Western Blot
Classification Monoclonal
Clone SR1313
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias DKFZp686C2056, DNA helicase, RecQ-like type 3, EC 3.1, EC 3.6.1, EC 3.6.4.12, Exonuclease WRN, RecQ protein-like 2, RecQ3, RECQ3RECQL3, RECQL2Werner syndrome ATP-dependent helicase, Werner syndrome, Werner syndrome, RecQ helicase-like
Host Species Rabbit
Immunogen A synthesized peptide derived from human WRN (Uniprot #: Q14191)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7486
Target Species Human
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.