missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xanthinol Nicotinate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Xanthinol Nicotinate
Specifications
Specifications
| Chemical Name or Material | Xanthinol Nicotinate |
| CAS | 437-74-1 |
| Analyte or Component Names | Xanthinol Nicotinate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H21 N5 O4 . C6 H5 N O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 3-Pyridinecarboxylic acid, compd. with 3,7-dihydro-7-[2-hydroxy-3-[(2-hydroxyethyl)methylamino]propyl]-1,3-dimethyl-1 H-purine-2,6-dione (1:1), Nicotinic acid, compd. with 7-[2-hydroxy-3-[(2-hydroxyethyl)methylamino]propyl]theophylline (1:1) (8 CI), Theophylline, 7-[2-hydroxy-3-[(2-hydroxyethyl)methylamino]propyl]-, nicotinate (6 CI), 1 H-Purine-2,6-dione, 3,7-dihydro-7-[2-hydroxy-3-[(2-hydroxyethyl)methylamino]propyl]-1,3-dimethyl-, mono-3-pyridinecarboxylate (salt) (9 CI), Theophylline, 7-[2-hydroxy-3-[(2-hydroxyethyl)methylamino]propyl]-, mononicotinate (salt) (8 CI), SK 331 A, SK 331 A, SK 331 A, SK 331 A, SK 331 A, SK 331 A, SK 331 A, SK 331 A, SK 331 A, SK 331 A, SK 331 A, SK 331 A, SK 331 A, SK 331 A, SK 331 A, SK 331 A, SK 331 A, SK 331 A, SK 331 A, SK 331 A, SK 331 A |
| InChI Formula | InChI=1 S/C13H21N5O4.C6H5NO2/c1-15(4-5-19)6-9(20)7-18-8-14-11-10(18)12(21)17(3)13(22)16(11)2;8-6(9)5-2-1-3-7-4-5/h8-9,19-20 H,4-7H2,1-3H3;1-4 H,(H,8,9) |
| SMILES | CN(CCO)CC(O)Cn1cnc2N(C)C(=O)N(C)C(=O)c12.OC(=O)c3cccnc3 |
| IUPAC Name | 7-[2-hydroxy-3-[2-hydroxyethyl(methyl)amino]propyl]-1,3-dimethylpurine-2,6-dione;pyridine-3-carboxylic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction