missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XKR6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68822
This item is not returnable.
View return policy
Description
XKR6 Polyclonal antibody specifically detects XKR6 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| XKR6 | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| C8orf5, C8orf7, FLJ31557, Transmembrane protein C8orf5, X Kell blood group precursor-related family, member 6, XK, Kell blood group complex subunit-related family, member 6, XK-related protein 6, XRG6 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: FVDRRLRRTINILQYVTPTAVGIRYRDGPLLYELLQYESSL | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 286046 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction