missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XRCC2 Mouse, anti-Human, CoraLite 555, Clone: 3D9A1, Proteintech
Mouse Monoclonal Antibody
145.00 € - 450.00 €
Specifications
| Antigen | XRCC2 |
|---|---|
| Clone | 3D9A1 |
| Concentration | 1000 μg/mL |
| Applications | Flow Cytometry (Intracellular Staining) |
| Conjugate | CoraLite 555 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18005135
|
Proteintech
CL555-66652-100UL |
100 μL |
450.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18005485
|
Proteintech
CL555-66652-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The XRCC2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human XRCC2. This antibody recognizes human, mouse antigen. The XRCC2 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™555.
Specifications
| XRCC2 | |
| 1000 μg/mL | |
| CoraLite 555 | |
| Mouse | |
| 7516 | |
| IgG2b | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 3D9A1 | |
| Flow Cytometry (Intracellular Staining) | |
| Liquid | |
| Human, Mouse | |
| DKFZp781P0919, DNA repair protein XRCC2, XRCC2 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title