missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YAP1 Antibody (1A12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-22117
This item is not returnable.
View return policy
Description
YAP1 Monoclonal specifically detects YAP1 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated, Knockdown Validated.
Specifications
| YAP1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:2000, Simple Western 20 ug/mL, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-5 ug/mL, Immunoprecipitation reported in scientific literature (PMID 28230103), Immunohistochemistry-Paraffin 1:200-1:1000, Knockout Validated, KnockDown Validated reported in scientific literature (PMID 28230103) | |
| 65 kDa Yes-associated protein, COB1, Protein Yorkie Homolog, Transcriptional Coactivator YAP1, YAP, YAP1-2gamma, YAP2, YAP2L, YAP65, YAP65yes-associated protein 2, Yes Associated Protein, Yes-Associated Protein, Yes-associated protein 1, Yes-associated protein 1, 65kDa, yes-associated protein beta, yes-associated protein delta, Yes-Associated Protein YAP65 Homolog, YKI, yorkie homolog | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Precipitation |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay, KnockDown | |
| 1A12 | |
| Unconjugated | |
| P46937 | |
| YAP1 | |
| This YAP1 antibody (1A12) was developed against recombinant human YAP1 protein expressed in E. coli. [UniProt# P46937] | |
| 0.1 mL | |
| Breast Cancer, Cancer, Chromatin Research, Core ESC Like Genes, Signal Transduction, Stem Cell Markers, Transcription Factors and Regulators, Tyrosine Kinases | |
| 10413 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction