missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YAP1 Antibody (1A12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
168.00 € - 474.00 €
Specifications
| Antigen | YAP1 |
|---|---|
| Clone | 1A12 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Simple Western 20 ug/mL, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-5 ug/mL, Immunoprecipitation reported in scientific literature (PMID 28230103), Immunohistochemistry-Paraffin 1:200-1:1000, Knockout Validated, KnockDown Validated reported in scientific literature (PMID 28230103) |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay, KnockDown |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18130702
|
Novus Biologicals
NBP2-22117SS |
0.025 mL |
168.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18757733
|
Novus Biologicals
NBP2-22117 |
0.1 mL |
474.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
YAP1 Monoclonal specifically detects YAP1 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated, Knockdown Validated.Specifications
| YAP1 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay, KnockDown | |
| Unconjugated | |
| Mouse | |
| Breast Cancer, Cancer, Chromatin Research, Core ESC Like Genes, Signal Transduction, Stem Cell Markers, Transcription Factors and Regulators, Tyrosine Kinases | |
| P46937 | |
| 10413 | |
| This YAP1 antibody (1A12) was developed against recombinant human YAP1 protein expressed in E. coli. [UniProt# P46937] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1A12 | |
| Western Blot 1:500-1:2000, Simple Western 20 ug/mL, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-5 ug/mL, Immunoprecipitation reported in scientific literature (PMID 28230103), Immunohistochemistry-Paraffin 1:200-1:1000, Knockout Validated, KnockDown Validated reported in scientific literature (PMID 28230103) | |
| Monoclonal | |
| Precipitation | |
| RUO | |
| Human | |
| 65 kDa Yes-associated protein, COB1, Protein Yorkie Homolog, Transcriptional Coactivator YAP1, YAP, YAP1-2gamma, YAP2, YAP2L, YAP65, YAP65yes-associated protein 2, Yes Associated Protein, Yes-Associated Protein, Yes-associated protein 1, Yes-associated protein 1, 65kDa, yes-associated protein beta, yes-associated protein delta, Yes-Associated Protein YAP65 Homolog, YKI, yorkie homolog | |
| YAP1 | |
| IgG1 | |
| Ammonium sulfate precipitation |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title