missing translation for 'onlineSavingsMsg'
Learn More

YTHDC1 Antibody - BSA Free, Novus Biologicals™

Product Code. 30556782 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30556782 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30556782 Supplier Novus Biologicals Supplier No. NBP345528

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

YTHDC1 Polyclonal antibody specifically detects YTHDC1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen YTHDC1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:200-1:1000, ELISA
Formulation PBS, 50% Glycerol, pH (7.3)
Gene Alias KIAA1966splicing factor YT521-B, Putative splicing factor YT521, YT521-B, YT521YTH domain-containing protein 1, YTH domain containing 1
Host Species Rabbit
Immunogen YTH domain containing 1. (Uniprot# Q96MU7)
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91746
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.