missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZAP70 (Chronic Lymphocytic Leukemia Marker) Mouse, Clone: ZAP70/2035, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 7535-MSM5-P1
Additional Details : Weight : 0.01000kg
Specifications
ZAP70 (Chronic Lymphocytic Leukemia Marker) | |
Cell Surface and Cytoplasmic | |
ZAP70/2035 | |
Unconjugated | |
Mouse | |
70kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
7535 | |
IgG1 κ |
Immunohistochemistry, ELISA | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant fragment of human ZAP70 protein (apprx. aa 247-382) | |
Jurkat cells. Tonsil or lymph node. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |