missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZAP70 (Chronic Lymphocytic Leukemia Marker) Mouse, Clone: ZAP70/2035, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 485.00 €
Specifications
Antigen | ZAP70 (Chronic Lymphocytic Leukemia Marker) |
---|---|
Clone | ZAP70/2035 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15991098
|
EnquireBio
7535-MSM5-P1 |
100 μg |
485.00 €
100µg |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15981098
|
EnquireBio
7535-MSM5-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
ZAP70 (Chronic Lymphocytic Leukemia Marker) | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant fragment of human ZAP70 protein (apprx. aa 247-382) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
70kDa |
ZAP70/2035 | |
Immunohistochemistry, ELISA | |
Unconjugated | |
RUO | |
7535 | |
IgG1 κ | |
Protein A/G purified from Bioreactor concentrate. | |
Cell Surface and Cytoplasmic | |
Jurkat cells. Tonsil or lymph node. |