missing translation for 'onlineSavingsMsg'
Learn More

ZBTB22 Antibody [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30582487 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30582487 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30582487 Supplier Novus Biologicals Supplier No. NBP345521MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZBTB22 Polyclonal antibody specifically detects ZBTB22 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB22
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias BING1ZNF297, fru, fruitless, Protein BING1, ZBTB22AZNF297A, zinc finger and BTB domain containing 22, zinc finger and BTB domain-containing protein 22, Zinc finger protein 297Zinc finger and BTB domain-containing protein 22A
Host Species Rabbit
Immunogen zinc finger and BTB domain containing 22. (Uniprot# O15209)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9278
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.