missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZDHHC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-13541
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
ZDHHC2 Polyclonal specifically detects ZDHHC2 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.
Spezifikation
| ZDHHC2 | |
| Polyclonal | |
| Western Blot Reactivity reported in (PMID: 26214373)., Simple Western 1:20, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| DHHC2, DHHC-2, EC 2.3.1, EC 2.3.1.-, palmitoyltransferase ZDHHC2, Ream, REC, Reduced expression associated with metastasis protein, Reduced expression in cancer protein, Zinc finger DHHC domain-containing protein 2, Zinc finger protein 372, zinc finger, DHHC domain containing 2, zinc finger, DHHC-type containing 2, ZNF372rec | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 51201 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ZDHHC2 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: KEFHLSYAEKDLLEREPRGEAHQEVLRRAAKDLPIYTRTMSGAIRYCDRCQLI | |
| 0.1 mL | |
| Primary | |
| Specificity of human ZDHHC2 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur