missing translation for 'onlineSavingsMsg'
Learn More

ZEB2 Antibody (CL13467) [Alexa Fluor™ 647], Novus Biologicals™

Product Code. 30297363 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30297363 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30297363 Supplier Novus Biologicals Supplier No. NBP324559AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZEB2 Monoclonal antibody specifically detects ZEB2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZEB2
Applications Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone CL13467
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias KIAA0569FLJ42816, SIP1HSPC082, Smad-interacting protein 1, SMADIP1ZFHX1BSIP-1, ZFX1B, zinc finger E-box binding homeobox 2, zinc finger E-box-binding homeobox 2, zinc finger homeobox 1b, Zinc finger homeobox protein 1b
Host Species Mouse
Immunogen This antibody was developed using a recombinant protein derived from O60315, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9839
Target Species Human
Content And Storage Store at 4⪚C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.