missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFP91 Antibody (7G11H2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61854-0.025ml
This item is not returnable.
View return policy
Description
ZFP91 Monoclonal specifically detects ZFP91 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ZFP91 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| FLJ57065, ZFP-91, zinc finger protein 91 homolog (mouse), zinc finger protein homologous to Zfp91 in mouse | |
| Mouse | |
| 82.5 kDa | |
| 0.025 mL | |
| 80829 | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry | |
| 7G11H2 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| ZFP91 | |
| Purified recombinant fragment of human ZFP91 (AA: 162-304) expressed in E. Coli. | |
| Protein G purified | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction