missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFP91 Antibody (7G11H2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
203.00 € - 552.00 €
Specifications
| Antigen | ZFP91 |
|---|---|
| Clone | 7G11H2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18298781
|
Novus Biologicals
NBP2-61854 |
0.1 mL |
552.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18661969
|
Novus Biologicals
NBP2-61854-0.025ml |
0.025 mL |
203.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZFP91 Monoclonal specifically detects ZFP91 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ZFP91 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| FLJ57065, ZFP-91, zinc finger protein 91 homolog (mouse), zinc finger protein homologous to Zfp91 in mouse | |
| ZFP91 | |
| IgG1 | |
| Protein G purified |
| 7G11H2 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Unconjugated | |
| Mouse | |
| 80829 | |
| Purified recombinant fragment of human ZFP91 (AA: 162-304) expressed in E. Coli. | |
| Primary | |
| 82.5 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title