missing translation for 'onlineSavingsMsg'
Learn More

ZFYVE28 Antibody (LST2/2426), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18575076 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18575076 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18575076 Supplier Novus Biologicals Supplier No. NBP308899AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZFYVE28 Monoclonal antibody specifically detects ZFYVE28 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFYVE28
Applications Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone LST2/2426
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias hLst2, KIAA1643, lateral signaling target protein 2 homolog, LST2, LYST2, MGC43699, MGC61592, Zinc finger FYVE domain-containing protein 28, zinc finger, FYVE domain containing 28
Host Species Mouse
Immunogen Recombinant full-length human ZFYVE28 protein (Uniprot: Q9H9)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57732
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.