missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zidovudine EP Impurity B, Mikromol™
Description
Zidovudine EP Impurity B
Specifications
Specifications
| Chemical Name or Material | 1-(3-Chloro-2,3-dideoxy-Beta-D-erythro-pentofuranosyl)-5-methylpyrimidine-2,4(1H,3H)-dione |
| CAS | 25526-94-7 |
| Analyte or Component Names | 1-(3-Chloro-2,3-dideoxy-Beta-D-erythro-pentofuranosyl)-5-methylpyrimidine-2,4(1H,3H)-dione |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H13 Cl N2 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1-(3-Chloro-2,3-dideoxy-β-D-erythro-pentofuranosyl)-5-methylpyrimidine-2,4(1H,3H)-dione, 1-(3-Chloro-2,3-dideoxy-beta-D-erythro-pentofuranosyl)-5-methylpyrimidine-2,4(1H,3H)-dione, Zidovudine Imp. B (EP), 892U81, GW 297366X, USP Zidovudine Related Compound B, Ph Eur Zidovudine Impurity B, 3-chloro-3-deoxythymidine 892U81, 3'-chloro-3'-deoxythymidine, Zidovudine Related Compound B, Zidovudine Impurity B |
| InChI Formula | InChI=1S/C10H13ClN2O4/c1-5-3-13(10(16)12-9(5)15)8-2-6(11)7(4-14)17-8/h3,6-8,14H,2,4H2,1H3,(H,12,15,16)/t6-,7+,8+/m0/s1 |
| SMILES | CC1=CN([C@H]2C[C@H](Cl)[C@@H](CO)O2)C(=O)NC1=O |
| IUPAC Name | 1-[(2R,4S,5R)-4-chloro-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?