Learn More
Invitrogen™ Zika Virus Envelope (strain Zika SPH2015) Recombinant Rabbit Monoclonal Antibody (033), HRP
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA560379
 View more versions of this product
Product Code. 30536104
  • 569.00 € / 100µg
Description
Zika Virus Envelope (strain Zika SPH2015) Recombinant Monoclonal Antibody for ELISA
As a member of the Flaviviridae family, ZIKV has a single positive sense RNA genome, which is initially translated as a single polyprotein which is then cleaved post-transitionally into three structural proteins (C, PrM or M, and E) and seven nonstructural (NS) proteins (NS1, NS2A, NS2B, NS3, NS4A, NS4B and NS5)(1). The glycosated NS1 forms a homodimer inside the cells and is necessary for viral replication and late in infection (2). NS1 is also secreted into the extracellular space as a hexameric lipoprotein particle, which assists the virus in immune evasion and pathogenesis by interacting with components from both innate and adaptive immune systems as well as other host factors (1). Reference 1) Song, H.; Qi, J.; Haywood, J.; Shi, Y.; Gao, G.F. (2016). Nat. Struct. Mol. Biol. 23, 456-458. 2) Suthar, M.S., Diamond, M.S. and Gale, M. Jr. (2013). Nat. Rev. Microbiol. 11, 115-128.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Zika Virus Envelope (strain Zika SPH2015)
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
Rabbit
Protein A
RUO
Virus
Antibody
IgG
ELISA
033
HRP
Zika Envelope
Recombinant Zika virus (ZIKV) (strain Zika SPH2015) ZIKV-E/Envelope protein (Domain III) protein (Val593-Lys699).
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles