missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZIP4H/Tex11 Polyclonal antibody specifically detects ZIP4H/Tex11 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | ZIP4H/Tex11 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | testis expressed 11, testis expressed sequence 11, testis-expressed sequence 11 protein, TGC1, TSGA3 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 701-940 of human ZIP4H/Tex11 (Q8IYF3). AFEQTMFLSRALEEIQTCNDIHNFLKQTGTFSNDSCEKLLLLYEFEVRAKLNDPLLESFLESVWELPHLETKTFETIAIIAMEKPAHYPLIALKALKKALLLYKKEEPIDISQYSKCMHNLVNLSVPDGASNVELCPLEEVWGYFEDALSHISRTKDYPEMEILWLMVKSWNTGVLMFSRSKYASAEKWCGLALRFLNHLTSFKESYETQMNMLYSQLVEALSNNKGPVFHEHGYWSKSD |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?