missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZMAT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12677-50ul
This item is not returnable.
View return policy
Description
ZMAT1 Polyclonal antibody specifically detects ZMAT1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| ZMAT1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 0.2-1uL/mg of uLysate, Immunohistochemistry-Paraffin 1:2500-1:10000 | |
| KIAA1789, zinc finger, matrin type 1, zinc finger, matrin-type 1 | |
| Produced in rabbits immunized with E. coli-derived Human ZMAT1 fragment | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 84460 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction