missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZMAT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
195.00 € - 322.00 €
Specifications
| Antigen | ZMAT1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 0.2-1uL/mg of uLysate, Immunohistochemistry-Paraffin 1:2500-1:10000 |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18695176
|
Novus Biologicals
NBP3-12677-100ul |
100 μg |
322.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18671678
|
Novus Biologicals
NBP3-12677-50ul |
50 μg |
195.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZMAT1 Polyclonal antibody specifically detects ZMAT1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| ZMAT1 | |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| KIAA1789, zinc finger, matrin type 1, zinc finger, matrin-type 1 | |
| Produced in rabbits immunized with E. coli-derived Human ZMAT1 fragment | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000, Immunoprecipitation 0.2-1uL/mg of uLysate, Immunohistochemistry-Paraffin 1:2500-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH7.0 | |
| 84460 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title