missing translation for 'onlineSavingsMsg'
Learn More

ZNF232 Antibody (PCRP-ZNF232-2B3), HRP, Novus Biologicals™

Product Code. 18682864 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18682864 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18682864 Supplier Novus Biologicals Supplier No. NBP324043H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF232 Monoclonal antibody specifically detects ZNF232 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF232
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PCRP-ZNF232-2B3
Conjugate HRP
Formulation PBS
Gene Alias zinc finger protein 500, Zinc finger protein with KRAB and SCAN domains 18, ZKSCAN18KIAA0557
Host Species Mouse
Immunogen Recombinant fragment (around aa29-109) of human ZNF232 protein
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7775
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.