missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF286B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00 € - 624.00 €
Specifications
| Antigen | ZNF286B |
|---|---|
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18661976
|
Novus Biologicals
NBP2-38115-25ul |
25 μL |
415.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18146728
|
Novus Biologicals
NBP2-38115 |
0.1 mL |
624.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNF286B Polyclonal specifically detects ZNF286B in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ZNF286B | |
| Polyclonal | |
| Rabbit | |
| Human | |
| P0CG31 | |
| 729288 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LSLTLTYYRTAFLLSTENEGNLHFQCPSDVETRPQS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Putative Zinc Finger Protein 286B, Zinc Finger 286C Pseudogene, Zinc Finger Protein 286B, Zinc Finger Protein 286-Like, Zinc Finger Protein 590, ZNF286C, ZNF286L, ZNF590 | |
| ZNF286B | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title