missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF541 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05636-20ul
This item is not returnable.
View return policy
Description
ZNF541 Polyclonal antibody specifically detects ZNF541 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| ZNF541 | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000 | |
| DKFZp434I1930, MGC119518, MGC119519, MGC119521, zinc finger protein 541 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of mouse ZNF541 (NP_001092747.1). MEPYSLGEEGALPSEGHLPSFSESQVLNCSDTLNRDLGPSTRDLLYAGLSGLDLDPSLSTSDMPSEVLEDNLDTLSLYSGKDSDSVKLLEEYADSESQTSLQDLGLGALKVPKEADEGGRATGSTRKGKRQHSSPQNPLLDCSLCGKVFSSASSLSKHYLTHSQERKHVCKVCSKAFKRQDHLTGHMLTHQKTKPFVCIEQGCSKSYCDYRSLRRHYEVQHGVCILKETPPEEEAYGDPTHNHDVANQPPPSGLRSLGPPEARSPGSVLPNRDLLRCIVSSIVHQKIPSPGPAVGPSDTE | |
| 20 μg | |
| Primary | |
| Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 84215 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction