missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF541 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
188.00 € - 463.00 €
Specifications
| Antigen | ZNF541 |
|---|---|
| Dilution | Western Blot 1:200 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18659757
|
Novus Biologicals
NBP3-05636-100ul |
100 μg |
463.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18628327
|
Novus Biologicals
NBP3-05636-20ul |
20 μg |
188.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNF541 Polyclonal antibody specifically detects ZNF541 in Mouse, Rat samples. It is validated for Western BlotSpecifications
| ZNF541 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat | |
| DKFZp434I1930, MGC119518, MGC119519, MGC119521, zinc finger protein 541 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of mouse ZNF541 (NP_001092747.1). MEPYSLGEEGALPSEGHLPSFSESQVLNCSDTLNRDLGPSTRDLLYAGLSGLDLDPSLSTSDMPSEVLEDNLDTLSLYSGKDSDSVKLLEEYADSESQTSLQDLGLGALKVPKEADEGGRATGSTRKGKRQHSSPQNPLLDCSLCGKVFSSASSLSKHYLTHSQERKHVCKVCSKAFKRQDHLTGHMLTHQKTKPFVCIEQGCSKSYCDYRSLRRHYEVQHGVCILKETPPEEEAYGDPTHNHDVANQPPPSGLRSLGPPEARSPGSVLPNRDLLRCIVSSIVHQKIPSPGPAVGPSDTE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:200 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 84215 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title