missing translation for 'onlineSavingsMsg'
Learn More

Zoxazolamine, LoGiCal

Product Code. 30461001
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30461001

Brand: LoGiCal LGCFOR3325.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zoxazolamine
CAS 61-80-3
Analyte or Component Names Zoxazolamine
Shipping Condition Room Temperature
Molecular Formula C7 H5 Cl N2 O
Certifications/Compliance ISO 9001
Synonym 2-Benzoxazolamine, 5-chloro-, Benzoxazole, 2-amino-5-chloro- (6CI,8CI), 2-Amino-5-chlorobenzoxazole, 5-Chloro-1,3-benzoxazol-2-amine, 5-Chloro-2-aminobenzoxazole, 5-Chlorobenzo[d]oxazol-2-amine, 5-Chlorobenzoxazol-2-amine, 5-Chlorobenzoxazol-2-ylamine, Deflexol, Flexilon, Flexin, McN-485, NSC 24995, Zoxamin, Zoxazolamine, Zoxine
InChI Formula InChI=1S/C7H5ClN2O/c8-4-1-2-6-5(3-4)10-7(9)11-6/h1-3H,(H2,9,10)
Solution Type Neat
SMILES Nc1oc2ccc(Cl)cc2n1
IUPAC Name 5-chloro-1,3-benzoxazol-2-amine
Industry Type Clinical Testing
Molecular Weight (g/mol) 168.58
Quantity 10 mg
Formula Weight 168.009 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.