missing translation for 'onlineSavingsMsg'
Learn More

ZSCAN4 Antibody [DyLight 680], Novus Biologicals Biologicals™

Product Code. 30501944 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30501944 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30501944 Supplier Novus Biologicals Supplier No. NBP335200FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZSCAN4 Polyclonal antibody specifically detects ZSCAN4 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZSCAN4
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias FLJ35105, MGC126787, zinc finger and SCAN domain containing 4, Zinc finger protein 494MGC126789, ZNF494zinc finger and SCAN domain-containing protein 4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-227 of human ZSCAN4 (NP_689890.1).,, Sequence:, MALDLRTIFQCEPSENNLGSENSAFQQSQGPAVQREEGISEFSRMVLNSFQDSNNSYARQELQRLYRIFHSWLQPEKHSKDEIISLLVLEQFMIGGHCNDKASVKEKWKSSGKNLERFIEDLTDDSINPPALVHVHMQGQEALFSEDMPLRDVIVHLTKQVNAQTTREANMGTPSQTSQDTSLETGQGYEDEQDGWNSSSKTTRVNENITNQGNQIVSLIIIQEENGPRPEEGGVSSDNPYNSKRAELVTARSQEGSINGITFQGVPMVMGAGCISQPEQSSPESALTHQSNEGNSTCEVHQKGSHGVQK
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201516
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.