missing translation for 'onlineSavingsMsg'
Learn More

ZSCAN4 Antibody [Janelia Fluor« 549], R&D Systems™

Product Code. 30498473 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30498473 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30498473 Supplier Novus Biologicals Supplier No. NBP335200JF549
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

ZSCAN4 Polyclonal antibody specifically detects ZSCAN4 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen ZSCAN4
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias FLJ35105, MGC126787, zinc finger and SCAN domain containing 4, Zinc finger protein 494MGC126789, ZNF494zinc finger and SCAN domain-containing protein 4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-227 of human ZSCAN4 (NP_689890.1).,, Sequence:, MALDLRTIFQCEPSENNLGSENSAFQQSQGPAVQREEGISEFSRMVLNSFQDSNNSYARQELQRLYRIFHSWLQPEKHSKDEIISLLVLEQFMIGGHCNDKASVKEKWKSSGKNLERFIEDLTDDSINPPALVHVHMQGQEALFSEDMPLRDVIVHLTKQVNAQTTREANMGTPSQTSQDTSLETGQGYEDEQDGWNSSSKTTRVNENITNQGNQIVSLIIIQEENGPRPEEGGVSSDNPYNSKRAELVTARSQEGSINGITFQGVPMVMGAGCISQPEQSSPESALTHQSNEGNSTCEVHQKGSHGVQK
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201516
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.