missing translation for 'onlineSavingsMsg'
Learn More

Lgr5/GPR49 Antibody (803420) [DyLight 650], R&D Systems™

Product Code. 18621700 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18621700 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18621700 Supplier Novus Biologicals Supplier No. FAB8240W
  • ONLINE EXCLUSIVE PRICE
    783.70 €
    667.80 € / 0.1mL
    Save 115.90 €
    14% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rat Monoclonal Antibody

Lgr5/GPR49 Monoclonal specifically detects Lgr5/GPR49 in Mouse samples. It is validated for Agonist Activity, CyTOF, Flow Cytometry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lgr5/GPR49
Applications Agonist Activity, CyTOF, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 803420
Conjugate DyLight 650
Dilution Flow Cytometry, Agonist Activity, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias FEX, G protein-coupled receptor 49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38
Host Species Rat
Immunogen E. coli-derived recombinant mouse Lgr5/GPR49, Lys375-Pro547, Accession # Q9Z1P4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Neuroscience, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8549
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.