CAS RN 133099-07-7
CAS RN 133099-07-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Darifenacin Hydrobromide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Darifenacin hydrobromide, MedChemExpress
MedChemExpress Darifenacin hydrobromide (UK-88525 hydrobromide) is a selective M3 muscarinic receptor antagonist with pKi of 8.9.