accessibility menu, dialog, popup

UserName

CH3NONN

CAS RN 138401-24-8

IUPAC Name: 4'-({2-butyl-4-oxo-1,3-diazaspiro[4.4]non-1-en-3-yl}methyl)-[1,1'-biphenyl]-2-carbonitrile
Synonyms: 4'-({2-butyl-4-oxo-1,3-diazaspiro[4.4]non-1-en-3-yl}methyl)-[1,1'-biphenyl]-2-carbonitrile
Molecular Weight (g/mol): 385.51
Molecular Formula: C25H27N3O
InChi Key: KWEQEHOPDHARIA-UHFFFAOYSA-N
SMILES: CCCCC1=NC2(CCCC2)C(=O)N1CC1=CC=C(C=C1)C1=CC=CC=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results