CAS RN 151262-57-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(2RS)-3-[(4-Fluorophenyl)sulfonyl]-2-hydroxy-2-methylpropanoic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
3-[(4-Fluorophenyl)sulfonyl]-2-hydroxy-2-methylpropanoic Acid, TRC
CAS: 151262-57-6 Molecular Formula: C10H11FO5S Molecular Weight (g/mol): 262.25 Synonym: Propanoic acid, 3-[(4-fluorophenyl)sulfonyl]-2-hydroxy-2-methyl-,3-[(4-Fluorophenyl)sulfonyl]-2-hydroxy-2-methylpropanoic acid IUPAC Name: 3-(4-fluorophenyl)sulfonyl-2-hydroxy-2-methylpropanoic acid SMILES: CC(O)(CS(=O)(=O)c1ccc(F)cc1)C(=O)O