accessibility menu, dialog, popup

UserName

NOONOBr

CAS RN 204326-43-2

IUPAC Name: 12-bromo-4,8-dioxa-2,5-diazatricyclo[7.4.0.0²,⁶]trideca-1(9),5,10,12-tetraen-3-one
Synonyms: 12-bromo-4,8-dioxa-2,5-diazatricyclo[7.4.0.0²,⁶]trideca-1(9),5,10,12-tetraen-3-one
Molecular Weight (g/mol): 269.05
Molecular Formula: C9H5BrN2O3
InChi Key: MUDRLQRJCGJJTB-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(OCC3=NOC(=O)N23)C=C1
Quantity 
  • (1)
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results