accessibility menu, dialog, popup

UserName

CAS RN 22117-85-7

H3COOOSNH2OO

CAS RN 22117-85-7

IUPAC Name: 2-methoxy-5-sulfamoylbenzoate
Synonyms: 2-methoxy-5-sulfamoylbenzoate
Molecular Weight (g/mol): 230.21
Molecular Formula: C8H8NO5S
InChi Key: SQAILWDRVDGLGY-UHFFFAOYSA-M
SMILES: COC1=CC=C(C=C1C([O-])=O)S(N)(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results