accessibility menu, dialog, popup

UserName

CAS RN 22204-24-6

H3CNNSOHOHOOHOHO(E)

CAS RN 22204-24-6

IUPAC Name: 1-methyl-2-[(1E)-2-(thiophen-2-yl)ethenyl]-1,4,5,6-tetrahydropyrimidine; 4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid
Synonyms: pyrantel pamoate
Molecular Weight (g/mol): 594.68
Molecular Formula: C34H30N2O6S
InChi Key: AQXXZDYPVDOQEE-MXDQRGINSA-N
SMILES: CN1CCCN=C1\C=C\C1=CC=CS1.OC(=O)C1=C(O)C(CC2=C3C=CC=CC3=CC(C(O)=O)=C2O)=C2C=CC=CC2=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results