accessibility menu, dialog, popup

UserName

OOON(S)(R)(S)(S)(R)CH3CH3(S)H3CHHH

CAS RN 2517964-85-9

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results