missing translation for 'onlineSavingsMsg'
Learn More

Abiraterone Acetate N-Oxide, TRC

Product Code. 18071630
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18071630 25 mg 25mg
17909749 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18071630 Supplier Toronto Research Chemicals Supplier No. TRCA10851025MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2517964-85-9
Molecular Formula C26 H33 N O3
Molecular Weight (g/mol) 407.55
Synonym Abiraterone Acetate N-Oxide, Androsta-5,16-dien-3-ol, 17-(1-oxido-3-pyridinyl)-, 3-acetate, (3β)- (ACI), Abiraterone Acetate N-Oxide, (3β)-17-(3-Pyridinyl)androsta-5,16-dien-3-ol Acetate (Ester) N-Oxide, CB 7630 N-Oxide;
IUPAC Name [(3S,8R,9S,10R,13S,14S)-10,13-dimethyl-17-(1-oxidopyridin-1-ium-3-yl)-2,3,4,7,8,9,11,12,14,15-decahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate
SMILES CC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@@]4(C)[C@@H](CC=C4c5ccc[n+]([O-])c5)[C@@H]3CC=C2C1

Specifications

Analyte or Component Names Abiraterone Acetate N-Oxide
Shipping Condition Room Temperature
InChI Formula InChI=1S/C26H33NO3/c1-17(28)30-20-10-12-25(2)19(15-20)6-7-21-23-9-8-22(18-5-4-14-27(29)16-18)26(23,3)13-11-24(21)25/h4-6,8,14,16,20-21,23-24H,7,9-13,15H2,1-3H3/t20-,21-,23-,24-,25-,26+/m0/s1
Solution Type Neat
Industry Type Forensics
Quantity 25 mg
Formula Weight 407.246
Chemical Name or Material Abiraterone Acetate N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.