CAS RN 328898-40-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tildipirosin, MedChemExpress
MedChemExpress Tildipirosin, a long-acting macrolide, has antibiotic activity.