accessibility menu, dialog, popup

UserName

CH3ONOClCH3CH3

CAS RN 34256-82-1

IUPAC Name: 2-chloro-N-(ethoxymethyl)-N-(2-ethyl-6-methylphenyl)acetamide
Synonyms: acetochlor harness acenit azetochlor epa pesticide chemical code 121601 erunit acetochlore caswell no. 003B nevirex C14H20ClNO2
Molecular Weight (g/mol): 269.77
Molecular Formula: C14H20ClNO2
InChi Key: VTNQPKFIQCLBDU-UHFFFAOYSA-N
SMILES: CCOCN(C(=O)CCl)C1=C(C)C=CC=C1CC
Molecular Weight (g/mol) 
  • (8)
  • (4)
  • (1)
Solvent or Matrix 
  • (3)
  • (2)
  • (2)
  • (3)
Concentration 
  • (4)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (7)
  • (1)
  • (1)
  • (3)
  • (8)
  • (4)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)