CAS RN 36321-43-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(2-Aminoethyl)-1-naphthylacetamide, TRC
CAS: 36321-43-4 Molecular Formula: C14 H16 N2 O Molecular Weight (g/mol): 228.29 Synonym: N-(2-Aminoethyl)-1-Naphthaleneacetamide,N-(2-Aminoethyl)-1-naphthylacetamide,Naphazoline Imp. A (EP),N-(2-Aminoethyl)-2-(naphthalen-1-yl)acetamide,Naphthylacetylethylenediamine IUPAC Name: N-(2-aminoethyl)-2-naphthalen-1-ylacetamide SMILES: NCCNC(=O)Cc1cccc2ccccc12