accessibility menu, dialog, popup

UserName

OHBrBrN

CAS RN 521-74-4

IUPAC Name: 5,7-dibromoquinolin-8-ol
Synonyms: broxyquinoline C9H5Br2NO broxyquinolinum dibromoksin fenilor colepur broxyquinolin dibromoxin broxykinolin dibromoxine
Molecular Weight (g/mol): 302.95
Molecular Formula: C9H5Br2NO
InChi Key: ZDASUJMDVPTNTF-UHFFFAOYSA-N
SMILES: OC1=C2N=CC=CC2=C(Br)C=C1Br
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results