accessibility menu, dialog, popup

UserName

CH3HO

CAS RN 5472-13-9

IUPAC Name: (2-methylphenyl)(phenyl)methanol
Synonyms: phenyl-O-tolylcarbinol
Molecular Weight (g/mol): 198.27
Molecular Formula: C14H14O
InChi Key: MXHXXJOHFRHBFB-UHFFFAOYNA-N
SMILES: CC1=CC=CC=C1C(O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results